Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267621.1 | NP_001254550.1 | 3 - 4 | 593 | 73 | |
NM_010278.2 | NP_034408.1 | 2 - 3 | 805 | 73 | ||
GenBank mRNA | AK089065.1 | - | 3 - 4 | 594 | 73 | |
U58972.1 | - | 2 - 3 | 780 | 73 | ||
U78312.1 | - | 2 - 3 | 805 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus Inventoried Probe spans exons |