Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010553.4 | NP_034683.1 | 6 - 7 | 996 | 80 | |
GenBank mRNA | AF077347.1 | - | 6 - 7 | 1005 | 80 | |
AK137541.1 | - | 6 - 7 | 957 | 80 | ||
BC120598.1 | - | 6 - 7 | 1003 | 40129521 | 80 | |
BC120600.1 | - | 6 - 7 | 1003 | 40129523 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |