Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 5 - 6 | 1022 | 97 | |
GenBank mRNA | AF084480.1 | - | 5 - 6 | 981 | 97 | |
AK017894.1 | - | 5 - 6 | 1023 | 97 | ||
AK036160.1 | - | 2 - 3 | 3168 | 97 | ||
AK137139.1 | - | 1 - 2 | 759 | 97 | ||
AK140172.1 | - | 5 - 6 | 810 | 97 | ||
AK141305.1 | - | 5 - 6 | 763 | 97 | ||
BC141399.1 | - | 5 - 6 | 925 | 9056012 | 97 | |
BC171975.1 | - | 5 - 6 | 925 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |