Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079844.2 | NP_001073313.1 | 5 - 6 | 1650 | 84 | |
NM_011821.3 | NP_035951.1 | 5 - 6 | 1650 | 84 | ||
GenBank mRNA | AF105268.1 | - | 6 - 7 | 1373 | 84 | |
AK132531.1 | - | 5 - 6 | 1387 | 84 | ||
AK149312.1 | - | 5 - 6 | 1417 | 84 | ||
AK164668.1 | - | 5 - 6 | 1652 | 84 | ||
BC023448.1 | - | 5 - 6 | 1064 | 5008374 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |