Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012012.4 | NP_036142.2 | 3 - 4 | 322 | 89 | |
GenBank mRNA | AJ238213.1 | - | 1 - 2 | 177 | 89 | |
AK028728.1 | - | 3 - 4 | 303 | 89 | ||
AK166425.1 | - | 3 - 4 | 298 | 89 | ||
BC006671.1 | - | 3 - 4 | 286 | 3499406 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |