Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012012.4 | NP_036142.2 | 12 - 13 | 2241 | 87 | |
GenBank mRNA | AJ238213.1 | - | 10 - 11 | 2096 | 87 | |
AK028728.1 | - | 12 - 13 | 2222 | 87 | ||
AK166425.1 | - | 12 - 13 | 2217 | 87 | ||
BC006671.1 | - | 12 - 13 | 2205 | 3499406 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |