Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016669.1 | NP_057878.1 | 6 - 7 | 852 | 68 | |
GenBank mRNA | AF039391.1 | - | 6 - 7 | 852 | 68 | |
AK138422.1 | - | 6 - 7 | 823 | 68 | ||
AK143116.1 | - | 6 - 7 | 842 | 68 | ||
AK163781.1 | - | 6 - 7 | 823 | 68 | ||
BC045159.1 | - | 6 - 7 | 813 | 6492374 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |