Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017468.3 | NP_059496.1 | 3 - 4 | 353 | 82 | |
GenBank mRNA | BC115848.1 | - | 3 - 4 | 265 | 40048943 | 82 |
U82698.1 | - | 3 - 4 | 354 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |