Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018782.2 | NP_061252.2 | 4 - 5 | 482 | 118 | |
GenBank mRNA | AB015595.1 | - | 4 - 5 | 486 | 118 | |
AF146525.1 | - | 2 - 3 | 159 | 118 | ||
AK137210.1 | - | 4 - 5 | 483 | 118 | ||
BC138537.1 | - | 1 - 2 | 109 | 8861557 | 118 | |
BC138540.1 | - | 1 - 2 | 109 | 8861560 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |