Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018818.2 | NP_061288.2 | 12 - 13 | 1560 | 92 | |
GenBank mRNA | AF218084.1 | - | 12 - 13 | 1569 | 92 | |
AK141830.1 | - | 11 - 12 | 1438 | 92 | ||
BC047461.1 | - | 12 - 13 | 1559 | 5067077 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |