Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019423.2 | NP_062296.1 | 5 - 6 | 550 | 61 | |
GenBank mRNA | AF170908.1 | - | 5 - 6 | 540 | 61 | |
AK014803.1 | - | 5 - 6 | 629 | 61 | ||
AK029855.1 | - | 5 - 6 | 531 | 61 | ||
AK044770.1 | - | 5 - 6 | 553 | 61 | ||
AK050397.1 | - | 5 - 6 | 553 | 61 | ||
BC098215.1 | - | 5 - 6 | 533 | 30617894 | 61 | |
KF987080.1 | - | 3 - 4 | 448 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |