Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019584.3 | NP_062530.2 | 10 - 11 | 1172 | 121 | |
GenBank mRNA | AF077302.2 | - | 9 - 10 | 1034 | 121 | |
AK028851.1 | - | 8 - 9 | 2614 | 121 | ||
AK031501.1 | - | 9 - 10 | 1799 | 121 | ||
AK032176.1 | - | 1173 | 121 | |||
AK050329.1 | - | 10 - 11 | 1173 | 121 | ||
AK077392.1 | - | 10 - 11 | 1259 | 121 | ||
AK083800.1 | - | 10 - 11 | 1162 | 121 | ||
AK151531.1 | - | 10 - 11 | 1139 | 121 | ||
BC005770.1 | - | 10 - 11 | 1153 | 2650059 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |