Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019757.1 | NP_062731.1 | 10 - 11 | 1151 | 75 | |
GenBank mRNA | AF083809.1 | - | 10 - 11 | 1151 | 75 | |
AK081476.1 | - | 10 - 11 | 1207 | 75 | ||
AK083686.1 | - | 1221 | 75 | |||
AK154822.1 | - | 10 - 11 | 1235 | 75 | ||
AK155370.1 | - | 10 - 11 | 1233 | 75 | ||
AK163951.1 | - | 10 - 11 | 1210 | 75 | ||
AK209977.1 | - | 1 - 2 | 73 | 75 | ||
BC006616.1 | - | 10 - 11 | 1232 | 3967009 | 75 | |
FJ786633.2 | - | 5 - 6 | 385 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |