Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159502.1 | NP_001152974.1 | 4 - 5 | 1656 | 90 | |
NM_020494.3 | NP_065240.2 | 4 - 5 | 1490 | 90 | ||
GenBank mRNA | AF214732.1 | - | 4 - 5 | 1470 | 90 | |
AK134339.1 | - | 4 - 5 | 1468 | 90 | ||
AK149810.1 | - | 4 - 5 | 1442 | 90 | ||
AK166360.1 | - | 4 - 5 | 1451 | 90 | ||
AK172557.1 | - | 4 - 5 | 1443 | 90 | ||
BC043721.1 | - | 4 - 5 | 1458 | 90 | ||
BC055048.1 | - | 4 - 5 | 1420 | 6484820 | 90 | |
BC055317.1 | - | 4 - 5 | 1656 | 5696084 | 90 | |
U46690.1 | - | 2 - 3 | 204 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |