Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253832.1 | NP_001240761.1 | 2 - 3 | 367 | 62 | |
NM_021438.4 | NP_067413.1 | 2 - 3 | 367 | 62 | ||
GenBank mRNA | AF270700.1 | - | 2 - 3 | 280 | 62 | |
AK008093.1 | - | 2 - 3 | 358 | 62 | ||
AK008132.1 | - | 2 - 3 | 353 | 62 | ||
AK012629.1 | - | 2 - 3 | 350 | 62 | ||
AK019399.1 | - | 2 - 3 | 338 | 62 | ||
AK028029.1 | - | 2 - 3 | 358 | 62 | ||
BC031716.1 | - | 2 - 3 | 337 | 3969355 | 62 | |
BC049284.1 | - | 2 - 3 | 345 | 6395407 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |