Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021890.3 | NP_068690.3 | 8 - 9 | 1178 | 105 | |
GenBank mRNA | AB041560.1 | - | 8 - 9 | 1120 | 105 | |
AK029318.1 | - | 8 - 9 | 1037 | 105 | ||
AK080414.1 | - | 8 - 9 | 1030 | 105 | ||
AK090042.1 | - | 8 - 9 | 1161 | 105 | ||
AK154534.1 | - | 8 - 9 | 1055 | 105 | ||
AK172509.1 | - | 8 - 9 | 1157 | 105 | ||
BC055950.1 | - | 8 - 9 | 859 | 105 | ||
BC071266.1 | - | 7 - 8 | 1166 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |