Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025903.2 | NP_080179.1 | 5 - 6 | 716 | 69 | |
GenBank mRNA | AK007687.1 | - | 5 - 6 | 717 | 69 | |
AK156625.1 | - | 4 - 5 | 792 | 69 | ||
AK165967.1 | - | 5 - 6 | 710 | 69 | ||
AK166130.1 | - | 5 - 6 | 784 | 69 | ||
AK166258.1 | - | 5 - 6 | 674 | 69 | ||
AK178654.1 | - | 2 - 3 | 187 | 69 | ||
AY298860.1 | - | 5 - 6 | 545 | 69 | ||
BC037434.1 | - | 5 - 6 | 672 | 2651172 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |