Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050014.1 | - | 4 - 5 | 396 | 65 | |
AK075771.1 | - | 4 - 5 | 383 | 65 | ||
AK075791.1 | - | 4 - 5 | 382 | 65 | ||
AK077853.1 | - | 4 - 5 | 314 | 65 | ||
AK149765.1 | - | 4 - 5 | 410 | 65 | ||
AK153951.1 | - | 4 - 5 | 301 | 65 | ||
AK161834.1 | - | 2 - 3 | 241 | 65 | ||
AK166476.1 | - | 4 - 5 | 410 | 65 | ||
BC002318.1 | - | 4 - 5 | 380 | 3593792 | 65 | |
CK625767.1 | - | 4 - 5 | 413 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |