Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133191.2 | NP_573454.2 | 5 - 6 | 422 | 56 | |
GenBank mRNA | BC005492.1 | - | 5 - 6 | 422 | 3487337 | 56 |
BC009098.1 | - | 5 - 6 | 480 | 3482190 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |