Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133743.1 | NP_598504.1 | 2 - 3 | 237 | 101 | |
GenBank mRNA | AK009998.1 | - | 2 - 3 | 257 | 101 | |
BC003749.1 | - | 2 - 3 | 239 | 101 | ||
BC016549.1 | - | 2 - 3 | 237 | 3585545 | 101 | |
BC094282.1 | - | 2 - 3 | 274 | 5029753 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |