Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133930.1 | NP_598691.1 | 5 - 6 | 969 | 86 | |
GenBank mRNA | AB009020.1 | - | 5 - 6 | 974 | 86 | |
AK077726.1 | - | 5 - 6 | 1114 | 86 | ||
AK155777.1 | - | 5 - 6 | 979 | 86 | ||
BC010804.1 | - | 5 - 6 | 969 | 4239786 | 86 | |
BC023893.1 | - | 5 - 6 | 980 | 5310111 | 86 | |
BC025932.1 | - | 5 - 6 | 875 | 5102571 | 86 | |
BC029065.1 | - | 5 - 6 | 1005 | 4935296 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |