Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138584.2 | NP_613050.1 | 6 - 7 | 807 | 89 | |
GenBank mRNA | AK009332.1 | - | 6 - 7 | 799 | 89 | |
AK011985.1 | - | 6 - 7 | 774 | 89 | ||
AK090017.1 | - | 6 - 7 | 795 | 89 | ||
AK149930.1 | - | 6 - 7 | 781 | 89 | ||
AK150215.1 | - | 6 - 7 | 808 | 89 | ||
AK153284.1 | - | 6 - 7 | 769 | 89 | ||
AK170666.1 | - | 6 - 7 | 800 | 89 | ||
AK187404.1 | - | 2 - 3 | 148 | 89 | ||
AK206667.1 | - | 2 - 3 | 147 | 89 | ||
AK209522.1 | - | 2 - 3 | 147 | 89 | ||
AK210536.1 | - | 2 - 3 | 147 | 89 | ||
AK211454.1 | - | 2 - 3 | 147 | 89 | ||
AK217468.1 | - | 2 - 3 | 147 | 89 | ||
AK218378.1 | - | 2 - 3 | 147 | 89 | ||
BC099434.1 | - | 5 - 6 | 572 | 30714184 | 89 | |
BC132377.1 | - | 6 - 7 | 784 | 40130654 | 89 | |
BC132403.1 | - | 6 - 7 | 784 | 40130680 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |