Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144884.2 | NP_659133.1 | 1 - 2 | 250 | 74 | |
GenBank mRNA | AJ298841.1 | - | 1 - 2 | 205 | 74 | |
AK038891.1 | - | 1 - 2 | 207 | 74 | ||
AK160361.1 | - | 1 - 2 | 206 | 74 | ||
BC017683.1 | - | 1 - 2 | 230 | 4238480 | 74 | |
BY724297.1 | - | 1 - 2 | 239 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |