Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145145.1 | NP_660127.1 | 8 - 9 | 794 | 55 | |
GenBank mRNA | AK134770.1 | - | 8 - 9 | 838 | 55 | |
AK153984.1 | - | 8 - 9 | 893 | 55 | ||
AY494857.1 | - | 8 - 9 | 854 | 55 | ||
BC027325.1 | - | 8 - 9 | 794 | 3981366 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |