Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145145.1 | NP_660127.1 | 18 - 19 | 1916 | 80 | |
GenBank mRNA | AK053889.1 | - | 17 - 18 | 2436 | 80 | |
AK134770.1 | - | 18 - 19 | 1960 | 80 | ||
AK153984.1 | - | 18 - 19 | 2015 | 80 | ||
AK197623.1 | - | 1 - 2 | 104 | 80 | ||
AY007238.1 | - | 18 - 19 | 1956 | 80 | ||
AY494857.1 | - | 18 - 19 | 1976 | 80 | ||
BC027325.1 | - | 18 - 19 | 1916 | 3981366 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |