Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145364.2 | NP_663339.1 | 7 - 8 | 927 | 100 | |
GenBank mRNA | BC018333.1 | - | 7 - 8 | 927 | 4162788 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |