Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145365.3 | NP_663340.1 | 9 - 10 | 1305 | 71 | |
GenBank mRNA | AF392874.1 | - | 7 - 8 | 956 | 71 | |
AK050186.1 | - | 7 - 8 | 947 | 71 | ||
AK077258.1 | - | 9 - 10 | 1306 | 71 | ||
BC010786.1 | - | 7 - 8 | 1000 | 4211480 | 71 | |
BC028820.1 | - | 7 - 8 | 977 | 5096773 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |