Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hkdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145419.1 | NP_663394.1 | 15 - 16 | 2313 | 88 | |
GenBank mRNA | AK145980.1 | - | 15 - 16 | 2292 | 88 | |
BC016235.1 | - | 15 - 16 | 2313 | 4504302 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |