Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145439.2 | NP_663414.3 | 16 - 17 | 2237 | 66 | |
GenBank mRNA | AK077671.1 | - | 14 - 15 | 1982 | 66 | |
AK090179.1 | - | 16 - 17 | 2192 | 66 | ||
AY236497.1 | - | 16 - 17 | 2136 | 66 | ||
AY263158.1 | - | 16 - 17 | 2162 | 66 | ||
BC004840.1 | - | 16 - 17 | 2162 | 2648849 | 66 | |
BC005510.1 | - | 9 - 10 | 1207 | 66 | ||
BC006956.1 | - | 14 - 15 | 1947 | 66 | ||
BC013502.1 | - | 7 - 8 | 964 | 4223599 | 66 | |
BC058195.1 | - | 16 - 17 | 2162 | 5356348 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |