Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029786.1 | - | 3 - 4 | 2030 | 85 | |
AK162464.1 | - | 2 - 3 | 2105 | 85 | ||
BC003865.1 | - | 2 - 3 | 232 | 85 | ||
BC072599.1 | - | 3 - 4 | 3449 | 85 | ||
BC079861.1 | - | 3 - 4 | 3436 | 6834767 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |