Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113416.1 | NP_001106887.1 | 7 - 8 | 719 | 76 | |
NM_145506.4 | NP_663481.2 | 7 - 8 | 719 | 76 | ||
GenBank mRNA | AK034676.1 | - | 7 - 8 | 641 | 76 | |
AK037024.1 | - | 7 - 8 | 675 | 76 | ||
AK040354.1 | - | 7 - 8 | 687 | 76 | ||
AK044959.1 | - | 7 - 8 | 657 | 76 | ||
AK044996.1 | - | 7 - 8 | 665 | 76 | ||
AK047523.1 | - | 7 - 8 | 646 | 76 | ||
AK079301.1 | - | 7 - 8 | 670 | 76 | ||
AK136125.1 | - | 7 - 8 | 674 | 76 | ||
AK161141.1 | - | 7 - 8 | 670 | 76 | ||
AK167449.1 | - | 7 - 8 | 719 | 76 | ||
AK173198.1 | - | 7 - 8 | 653 | 76 | ||
BC003937.1 | - | 7 - 8 | 704 | 3486543 | 76 | |
BC011476.1 | - | 7 - 8 | 653 | 4014009 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |