Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_001135151.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_001135151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_001135152.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_001135152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_144808.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
NM_144808.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018204.mm.1 |
TC1400002336.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_001135151.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_001135151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_001135152.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_001135152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_144808.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
NM_144808.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018203.mm.1 |
TC1400002336.mm.1 |
MTA TC |