Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144816.1 | NP_659065.1 | 5 - 6 | 790 | 115 | |
GenBank mRNA | AK140428.1 | - | 5 - 6 | 821 | 115 | |
BC021549.1 | - | 5 - 6 | 790 | 4501997 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |