Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291442.1 | NP_001278371.1 | 8 - 9 | 1086 | 84 | |
NM_001291443.1 | NP_001278372.1 | 9 - 10 | 1080 | 84 | ||
NM_144822.3 | NP_659071.1 | 8 - 9 | 1068 | 84 | ||
GenBank mRNA | AK046049.1 | - | 8 - 9 | 1051 | 84 | |
AK049521.1 | - | 8 - 9 | 1064 | 84 | ||
AK050113.1 | - | 8 - 9 | 1055 | 84 | ||
AK052485.1 | - | 8 - 9 | 1016 | 84 | ||
AK077605.1 | - | 8 - 9 | 1018 | 84 | ||
AK085553.1 | - | 8 - 9 | 987 | 84 | ||
BC018320.1 | - | 8 - 9 | 1009 | 4192966 | 84 | |
BC023022.1 | - | 9 - 10 | 1015 | 5367511 | 84 | |
BC023042.1 | - | 7 - 8 | 952 | 5357794 | 84 | |
BC026566.1 | - | 8 - 9 | 1019 | 4977083 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |