Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144874.4 | NP_659123.2 | 6 - 7 | 932 | 55 | |
GenBank mRNA | AK048099.1 | - | 6 - 7 | 914 | 55 | |
AK051792.1 | - | 6 - 7 | 831 | 55 | ||
AK141631.1 | - | 6 - 7 | 864 | 55 | ||
BC011509.1 | - | 6 - 7 | 892 | 3981950 | 55 | |
BC021498.1 | - | 6 - 7 | 852 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |