Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144896.4 | NP_659145.1 | 1 - 2 | 191 | 59 | |
GenBank mRNA | AK028193.1 | - | 1 - 2 | 190 | 59 | |
AK034702.1 | - | 1 - 2 | 192 | 59 | ||
AK053978.1 | - | 1 - 2 | 194 | 59 | ||
AK081800.1 | - | 1 - 2 | 195 | 59 | ||
AK134831.1 | - | 1 - 2 | 126 | 59 | ||
AK166231.1 | - | 1 - 2 | 202 | 59 | ||
BC005709.1 | - | 1 - 2 | 193 | 3157973 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |