Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163461.2 | NP_001156933.1 | 7 - 8 | 617 | 81 | |
NM_001163462.2 | NP_001156934.1 | 7 - 8 | 674 | 81 | ||
NM_001289569.1 | NP_001276498.1 | 7 - 8 | 631 | 81 | ||
NM_001289570.1 | NP_001276499.1 | 7 - 8 | 617 | 81 | ||
NM_144911.3 | NP_659160.2 | 7 - 8 | 617 | 81 | ||
GenBank mRNA | AK034418.1 | - | 7 - 8 | 633 | 81 | |
AK049889.1 | - | 7 - 8 | 674 | 81 | ||
AK145830.1 | - | 7 - 8 | 592 | 81 | ||
AK161901.1 | - | 7 - 8 | 598 | 81 | ||
AK166574.1 | - | 7 - 8 | 540 | 81 | ||
BC021895.1 | - | 7 - 8 | 578 | 3481916 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |