Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253692.1 | NP_001240621.1 | 7 - 8 | 802 | 72 | |
NM_144917.5 | NP_659166.1 | 8 - 9 | 833 | 72 | ||
GenBank mRNA | AK049155.1 | - | 8 - 9 | 763 | 72 | |
AK084010.1 | - | 7 - 8 | 733 | 72 | ||
AK164974.1 | - | 7 - 8 | 784 | 72 | ||
BC016193.1 | - | 6 - 7 | 501 | 4487200 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |