Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23146396 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2184304 |
BC020077.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2184304 |
NM_145549.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2015312 |
BC020077.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2015312 |
BC112327.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2015312 |
AK142188.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2015312 |
AK143777.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2015312 |
AK220422.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
267738 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829838 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47359 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260532 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491245 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
39925 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572236 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
509449 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
672456 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669136 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
787621 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817448 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174890 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
504885 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918141 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
713805 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426353 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
955166 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676177 |
NM_011282.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676177 |
U15443.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676177 |
X81650.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
133059 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342620 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648950 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576120 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894877 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
63795 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153924 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88791 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
36402 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11202 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142196 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P170634 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2184304 |
BC020077.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2184304 |
NM_145549.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2015312 |
BC020077.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2015312 |
BC112327.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2015312 |
AK142188.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2015312 |
AK143777.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2015312 |
AK220422.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146405 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146407 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146399 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146396 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146400 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146398 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146410 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146404 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146411 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146397 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146402 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23146414 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05333020 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05333019 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P170634 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P485275 |
BC020077.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P485275 |
BC112327.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P485275 |
AK142188.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P485275 |
AK220422.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P485275 |
NM_145549.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125717 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117329 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P111492 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101821 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P170634 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05333020 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05333019 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03595408 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03292012 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03595412 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03595405 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03261036 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03773264 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03595403 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03773267 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03595406 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03344971 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03420111 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23146396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146398 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23146444 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146497 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146476 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03420564 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146406 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146413 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146396 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146402 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146488 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04033972 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03491070 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03595425 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04018749 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03595405 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146410 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146489 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146493 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03595412 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146484 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146395 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03595403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146498 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23146404 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03990104 |
- |
- |