Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190343.1 | NP_001177272.1 | 5 - 6 | 682 | 100 | |
NM_001190344.1 | NP_001177273.1 | 6 - 7 | 768 | 100 | ||
NM_146014.3 | NP_666126.1 | 6 - 7 | 856 | 100 | ||
GenBank mRNA | AK155145.1 | - | 6 - 7 | 830 | 100 | |
AK166131.1 | - | 3 - 4 | 383 | 100 | ||
AK169727.1 | - | 6 - 7 | 770 | 100 | ||
AY442689.1 | - | 6 - 7 | 743 | 100 | ||
BC029157.1 | - | 6 - 7 | 787 | 4952288 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |