Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146047.2 | NP_666159.2 | 12 - 13 | 1522 | 83 | |
GenBank mRNA | AK032295.1 | - | 12 - 13 | 1486 | 83 | |
AK048357.1 | - | 12 - 13 | 1419 | 83 | ||
AK156203.1 | - | 12 - 13 | 1413 | 83 | ||
AK165939.1 | - | 12 - 13 | 1383 | 83 | ||
AK181076.1 | - | 4 - 5 | 244 | 83 | ||
BC026562.1 | - | 12 - 13 | 1223 | 5028264 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |