Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177796.1 | NP_001171267.1 | 7 - 8 | 1076 | 70 | |
NM_001177797.1 | NP_001171268.1 | 6 - 7 | 1022 | 70 | ||
NM_146102.2 | NP_666214.1 | 8 - 9 | 1128 | 70 | ||
GenBank mRNA | AK048649.1 | - | 8 - 9 | 782 | 70 | |
AK053222.1 | - | 10 - 11 | 1378 | 70 | ||
AK087449.1 | - | 7 - 8 | 712 | 70 | ||
AK220430.1 | - | 6 - 7 | 645 | 70 | ||
BC031515.1 | - | 7 - 8 | 678 | 3711746 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |