Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164357.1 | NP_001157829.1 | 7 - 8 | 1153 | 56 | |
NM_001164358.1 | NP_001157830.1 | 8 - 9 | 1408 | 56 | ||
NM_001290558.1 | NP_001277487.1 | 7 - 8 | 1022 | 56 | ||
NM_146118.3 | NP_666230.2 | 8 - 9 | 1189 | 56 | ||
GenBank mRNA | AK049392.1 | - | 7 - 8 | 1129 | 56 | |
AK082756.1 | - | 7 - 8 | 1110 | 56 | ||
AK122568.2 | - | 8 - 9 | 1373 | 56 | ||
AK132204.1 | - | 7 - 8 | 1107 | 56 | ||
BC019978.1 | - | 7 - 8 | 961 | 4239441 | 56 | |
BC022114.1 | - | 7 - 8 | 959 | 5098924 | 56 | |
BC037109.1 | - | 8 - 9 | 1149 | 5368306 | 56 | |
BC066998.1 | - | 8 - 9 | 1376 | 5703622 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |