Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198785.1 | NP_001185714.1 | 8 - 9 | 1063 | 72 | |
NM_001198787.1 | NP_001185716.1 | 8 - 9 | 835 | 72 | ||
NM_007377.4 | NP_031403.2 | 8 - 9 | 1346 | 72 | ||
GenBank mRNA | AB093253.2 | - | 8 - 9 | 1346 | 72 | |
AB288871.1 | - | 8 - 9 | 965 | 72 | ||
AF011908.1 | - | 8 - 9 | 802 | 72 | ||
AK147239.1 | - | 5 - 6 | 856 | 72 | ||
AK155100.1 | - | 8 - 9 | 837 | 72 | ||
AY236858.1 | - | 8 - 9 | 841 | 72 | ||
AY236859.1 | - | 8 - 9 | 868 | 72 | ||
BC042837.1 | - | 8 - 9 | 1368 | 72 | ||
BC052323.1 | - | 8 - 9 | 1368 | 72 | ||
BC080846.1 | - | 8 - 9 | 917 | 6856191 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |