Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024181.2 | NP_077143.2 | 14 - 15 | 1925 | 58 | |
GenBank mRNA | AF255459.1 | - | 14 - 15 | 1602 | 58 | |
AF314002.1 | - | 13 - 14 | 1438 | 58 | ||
AK004617.1 | - | 14 - 15 | 1680 | 58 | ||
AK028184.1 | - | 14 - 15 | 1666 | 58 | ||
AK128945.1 | - | 14 - 15 | 1679 | 58 | ||
AK165652.1 | - | 14 - 15 | 1682 | 58 | ||
AK166502.1 | - | 14 - 15 | 1636 | 58 | ||
AK169020.1 | - | 14 - 15 | 1692 | 58 | ||
BC002207.1 | - | 5 - 6 | 487 | 58 | ||
BC033461.1 | - | 14 - 15 | 1664 | 4010341 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |