Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293791.1 | NP_001280720.1 | 11 - 12 | 1472 | 82 | |
NM_001293792.1 | NP_001280721.1 | 11 - 12 | 1472 | 82 | ||
NM_027590.4 | NP_081866.1 | 11 - 12 | 1472 | 82 | ||
GenBank mRNA | AK014939.1 | - | 11 - 12 | 1448 | 82 | |
AK030974.1 | - | 11 - 12 | 1436 | 82 | ||
AK034498.1 | - | 11 - 12 | 1388 | 82 | ||
AK040574.1 | - | 11 - 12 | 1473 | 82 | ||
AK047093.1 | - | 11 - 12 | 1468 | 82 | ||
AK147743.1 | - | 11 - 12 | 1447 | 82 | ||
BC031923.1 | - | 11 - 12 | 1435 | 4486323 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |