Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134829.1 | NP_001128301.1 | 2 - 3 | 467 | 137 | |
NM_172266.3 | NP_758470.1 | 2 - 3 | 446 | 137 | ||
GenBank mRNA | AK133814.1 | - | 2 - 3 | 447 | 137 | |
AK172914.1 | - | 2 - 3 | 417 | 137 | ||
BC013667.1 | - | 2 - 3 | 355 | 4020003 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |