Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293689.1 | NP_001280618.1 | 2 - 3 | 790 | 72 | |
NM_172271.2 | NP_758475.1 | 2 - 3 | 790 | 72 | ||
NR_121629.1 | 1 - 2 | 377 | 72 | |||
GenBank mRNA | AK032262.1 | - | 1 - 2 | 167 | 72 | |
AK083807.1 | - | 2 - 3 | 691 | 72 | ||
AK085620.1 | - | 2 - 3 | 523 | 72 | ||
AK163113.1 | - | 2 - 3 | 794 | 72 | ||
AY155578.1 | - | 2 - 3 | 791 | 72 | ||
BC033610.1 | - | 2 - 3 | 415 | 72 | ||
BC141148.1 | - | 1 - 2 | 361 | 9055760 | 72 | |
BC171962.1 | - | 1 - 2 | 361 | 9054248 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |