Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172295.4 | NP_758499.3 | 2 - 3 | 579 | 77 | |
GenBank mRNA | AK029040.1 | - | 2 - 3 | 318 | 77 | |
AK161057.1 | - | 2 - 3 | 496 | 77 | ||
AK166928.1 | - | 329 | 77 | |||
AK172389.1 | - | 2 - 3 | 528 | 77 | ||
BC037703.1 | - | 2 - 3 | 516 | 4166596 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |