Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172437.3 | NP_766025.1 | 3 - 4 | 1130 | 116 | |
GenBank mRNA | AK019372.1 | - | 3 - 4 | 1122 | 116 | |
AK029027.1 | - | 3 - 4 | 1114 | 116 | ||
BC066798.1 | - | 2 - 3 | 1083 | 30469996 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |